Learn More
Thermo Scientific™ Valco™ Injector Model C6W
Catalog No. 03052441
Change view
Click to view available options
Type:
Injector/Switching Valve
Rotor Seal Valve
Volume (Metric) Sample:
100 μL
N/A
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Volume (Metric) Sample
03-052-441 Injector/Switching Valve 100 μL
03-170-308 Rotor Seal Valve N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-052-441 Supplier Thermo Scientific™ Supplier No. SL100CW
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. Available with manual or a microelectric actuator. Has 1/16 in. fittings and six 0.010 inch ports.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Actuation Microelectric or Manual
Volume (Metric) Sample 100 μL
Description Sample Injector Loop
Material Stainless Steel
Type Injector/Switching Valve
No. of Ports 6 Ports
Stator Material Stainless Steel
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.