Learn More
Leap PAL Parts VALCO REPLACEMENT ROTOR, VALCO
Supplier:  Leap PAL Parts KVAL.C520R4
Encompass
Valco Replacement Rotor, Valcon H, 4 position selector, 0.40mm bore, 1/16" for C55-2004I
Catalog No. NC2071421
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.