Learn More
MilliporeSigma™ Supelco™ Valco™ Unions Internal Reducing, 1/4 to 1/16 in., Bore Size 0.75 mm
Catalog No. 111012023 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-101-2023 1/Pk.
1 options
Catalog No. 11-101-2023 Supplier MilliporeSigma™ Supelco™ Supplier No. 58249
Add to Cart
Add to Cart
Stainless steel, unless noted otherwise. Nuts and ferrules included.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Description zero dead volume 0.75 mm (bore)
Material Stainless Steel
Valco is a registered trademark of Valco Instruments Co. Inc.