Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valeraldehyde, 96%

Catalog No. AAB213890B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB213890B 1000 g
AAB2138930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB213890B Supplier Thermo Scientific Chemicals Supplier No. B213890B
Add to Cart

CAS: 110-62-3 | C5H10O | 86.13 g/mol

It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application in flavorings, resin chemistry, and rubber accelerators. Valeraldehyde is used as a gaseous standard in the study of the sorptive loss pattern for volatile compounds. Valeraldehyde is used as an intermediate in industrial settings in the production of valeric acid and amyl alcohol.

Solubility
Soluble in water at 20°C, 13.5g/L. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-62-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00007026
InChI Key HGBOYTHUEUWSSQ-UHFFFAOYSA-N
Synonym valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde
PubChem CID 8063
ChEBI CHEBI:84069
IUPAC Name pentanal
SMILES CCCCC=O

Specifications

Melting Point -92°C
Density 0.81
Boiling Point 102°C to 103°C
Flash Point 8°C (46°F)
Odor Like Aldehyde
Refractive Index 1.395
Linear Formula CH3(CH2)3CHO
Quantity 1000 g
UN Number UN2058
Beilstein 1616304
Sensitivity Air Sensitive
Merck Index 14,9902
Solubility Information Soluble in water at 20°C,13.5g/L. Soluble in alcohol.
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material Valeraldehyde
Show More Show Less
Hazard Category H225-H317-H319-H332-H335
Hazard Statement GHS H Statement
H225-H318-H332-H335-H315
Highly flammable liquid and vapour.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VALERALDEHYDE
EINECSNumber 203-784-4
RTECSNumber YV3600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.