Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Valerenic acid | 3569-10-6 | MFCD00075694 | 99.0% | 234.33 g/mol | C15H22O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10352410MG

Encompass_Preferred

Valerenic acid is a sesquiterpenoid natural product that acts as a positive allosteric modulator of GABAA receptors and a partial agonist of 5-HT5A. It is used in pharmacology and neuroscience research to investigate GABAergic modulation, anxiolytic mechanisms, and antioxidant activity.

  • Purity 99.0%.
  • Available in 1 mg, 5 mg, 10 mg, 50 mg, and 100 mg pack sizes.
  • Molecular formula C15H22O2; molecular weight 234.33 g/mol.
  • Intended for in vitro and in vivo pharmacology studies of GABA receptor modulation and serotonergic pathways.
  • Typically prepared as DMSO stock solutions for assay use.

Catalog No. 50-002-45333


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.