Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valerenic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014018 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4018 1 mg
11-101-4019 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4018 Supplier MilliporeSigma Supelco Supplier No. 519641MG
Add to Cart
Add to Cart

Valerenic acid is a bioactive sesquiterpene and a natural product, isolated from Valeriana officinalis type of plant. It can find applications in the treatment of several dis-functions of the central nervous system, since it is potent modulator of the GABAA receptor.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 3569-10-6
Physical Form Neat
Linear Formula C15H22O2
MDL Number MFCD00075694
Synonym (2 E)-3-[(4 S,7 R,7 aR)-2,4,5,6,7,7 A-Hexadydro-3,7-dimethyl- 1 H-inden-4-yl]-2-methyl-2-propenoic acid
Type Analytical Standard
Quantity 1 mg
Formula Weight 234.33
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.