Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valeroin 97.0+%, TCI America™
SDP

Catalog No. V005525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V005525ML 25 mL
V00555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V005525ML Supplier TCI America Supplier No. V005525ML

Chemical Identifiers

CAS 6540-98-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00059663
InChI Key ORLDHCIQVZTBQQ-UHFFFAOYSA-N
Synonym 6-Hydroxy-5-decanone
PubChem CID 110894
IUPAC Name 6-hydroxydecan-5-one
SMILES CCCCC(C(=O)CCCC)O

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Valeroin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.