Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valerophenone, 99%

Catalog No. AAA1052514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1052514 25 g
AAA1052522 100 g
AAA1052536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1052514 Supplier Thermo Scientific Chemicals Supplier No. A1052514
Add to Cart

CAS: 1009-14-9 | C11H14O | 162.23 g/mol

Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-14-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009480
InChI Key XKGLSKVNOSHTAD-UHFFFAOYSA-N
Synonym valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci
PubChem CID 66093
ChEBI CHEBI:36812
IUPAC Name 1-phenylpentan-1-one
SMILES CCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point °C to 9°C
Density 0.978 g/mL
Boiling Point 244°C to 245°C
Flash Point 102°C (215°F)
Assay Percent Range 99%
Refractive Index 1.5143
Quantity 25 g
Beilstein 1907717
Solubility Information Insoluble in water.
Formula Weight 162.23
Percent Purity 99%
Chemical Name or Material Valerophenone
Show More Show Less
EINECSNumber 213-767-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.