Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valganciclovir hydrochloride, 97+%, Thermo Scientific Chemicals

Catalog No. AC461270010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC461270010 Supplier Thermo Scientific Chemicals Supplier No. 461270010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175865-59-5
Molecular Formula C14H23ClN6O5
Synonym valcyte tn

Specifications

Assay Percent Range 97% min. ( sum of two diastereomers) (HPLC)
Quantity 1 g
Formula Weight 390.82
Purity 97+%
Chemical Name or Material Valganciclovir hydrochloride
Hazard Category Reproductive toxicity (category 1B)
Hazard Statement May damage fertility or the unborn child

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.