Learn More
Sartorius iQue Screener Plus Validation Beads
Catalog No. NC1085263 Shop All Sartorius Products
Encompass
Perform daily quality control maintenance using iQue Screener Plus Validation Beads – easy to use and economic with two simple drops in 300 uL Qsol or Buffer.
Catalog No. NC1085263 Supplier Sartorius Supplier No. 91091
Perform daily quality control maintenance using iQue Screener Plus Validation Beads – easy to use and economic with two simple drops in 300 uL Qsol or Buffer.