Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Description
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
- Information on the validation process
- Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
- Information on pharmacopoeia recommended tests for specific instrument performance where applicable
- Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
Specifications
Specifications
| Type | Performance Verification Kit |
| For Use With (Equipment) | GENESYS 40-180, BioMate 160 Spectrophotometers |