Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valinomycin 96%, Thermo Scientific™

Catalog No. AC228880100
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC228880100 10 mg
AC228880500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228880100 Supplier Thermo Scientific Chemicals Supplier No. 228880100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.34
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-UYBNATROSA-N
Synonym valinomycin
PubChem CID 131668508
IUPAC Name (3S,6S,9R,12R,15S,18S,21R,24R,27S,30S,36R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nonakis(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexaazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)C(NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

Specifications

Melting Point 187°C to 191°C
Color White
Infrared Spectrum Authentic
Quantity 10 mg
Beilstein 27, IV, 9728
Merck Index 15, 10096
Specific Rotation +30° to +32° (20°C, 589nm) (c=1.6, benzene)
Formula Weight 1111.35
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material Valinomycin, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 217-896-6
RTECSNumber YV9468000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.