Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valproate Sodium, G-Biosciences
SCI_ED

Catalog No. S139770
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139770 25 g
S157238 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139770 Supplier G-Biosciences Supplier No. RC1130
Add to Cart
Add to Cart

Valproate sodium is a branched chain fatty acid with anticancer, neuroprotective, and anti-inflammatory properties.

It is also an inhibitor of histone deacetylase (HDAC) through promotion of transcriptionally active chromatin. This inhibition may regulate the neuroprotective effects of valproic acid.

Chemical Identifiers

Molecular Weight (g/mol) 166.19 g/mol

Specifications

Boiling Point 219.5°C
Quantity 25 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.