Learn More
Selleck Chemical LLC Valproic acid (VPA) 25mg 99-66-1 2-Propylvaleric Acid, Sodium valproate
SDP
Supplier:  Selleck Chemical LLC S394425MG
Encompass_Preferred
Valproic acid (VPA, 2-Propylvaleric Acid, Sodium valproate) is a fatty acid with anticonvulsant properties used in the treatment of epilepsy. It is also a histone deacetylase (HDAC) inhibitor and is under investigation for treatment of HIV and various cancers. Valproic acid (VPA) induces autophagy and mitophagy by upregulation of BNIP3 and mitochondrial biogenesis by upregulating PGC-1?. Valproic acid activates Notch-1 signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8061