Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Valproic Acid Monoclonal Antibody (VP10-234.5), Invitrogen™

Catalog No. p-7245316
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546166 100 μg
01-671-997 500 μg
PIMA546139 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546166 Supplier Invitrogen™ Supplier No. MA546166
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10767 detects Valproic Acid. MA1-10767 has been successfully used in ELISA applications. The MA1-10767 immunogen is Valproic acid, conjugated to KLH.

Valproic acid, valproate sodium, and divalproex belong to the group of medicines called anticonvulsants. They are used to control certain types of seizures in the treatment of epilepsy. Also Valproic acid relieves histone deacetylases (HDACs) dependent transcriptional repression and causes hyperacetylation of histones in cultured cells and in vivo. Valproic acid inhibits HDAC activity in vitro, most probably by binding to the catalytic center of HDACs. Most importantly, valproic acid induces differentiation of carcinoma cells, transformed hematopoietic progenitor cells and leukemic blasts from acute myeloid leukemia patients. More over, tumor growth and metastasis formation are significantly reduced in animal experiments. Therefore, valproic acid might serve as an effective drug for cancer therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Valproic Acid
Applications ELISA
Classification Monoclonal
Clone VP10-234.5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C8H16O2; Depakene; Depakine; Dipropylacetic acid
Host Species Mouse
Immunogen Valproic acid, conjugated to KLH.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.