Learn More
Valproic Acid Monoclonal Antibody (VP13-615.3), Invitrogen™
Description
MA1-10769 detects Valproic Acid. MA1-10769 has been successfully used in ELISA applications. The MA1-10769 immunogen is Valproic acid, conjugated to KLH.
Specifications
Specifications
| Antigen | Valproic Acid |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | VP13-615.3 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | C8H16O2; Depakene; Depakine; Dipropylacetic acid |
| Host Species | Mouse |
| Immunogen | Valproic acid, conjugated to KLH. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.