Learn More
International Enviroguard™ ValuGuard MP™ Lightweight Microporous Coveralls with Attached Hood and Boots
Catalog No. 17184044 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-044 X-Large
17-184-045 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-184-044 Supplier International Enviroguard™ Supplier No. 8119XL
Add to Cart
Add to Cart
Economical coveralls are made of lightweight microporous fabric that provides high moisture vapor transmission rate (HMVT) to help workers stay cool and prevent heat stress.
  • Fabric provides a high moisture vapor transmission rate (MVTR), helping you stay cooler to prevent heat stress
  • Coveralls protect against particulates, light liquid splashes, and sprays in nonhazardous work environments
  • Attached hood and boots offer full-body protection from head to toe
  • Keep productivity high and risk level low when wearing comfortable ValuGuard MP Coveralls
  • Secure, elastic wrists help keep sleeves in place to prevent skin exposure
  • Standard zipper and set-in sleeves
  • Environmentally friendly; can be recycled after use
Specifications
Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Tears, Liquid, Particle
Latex Free Yes
Model 8119
No. of Pockets None