Learn More
International Enviroguard™ ValuGuard MP™ Lightweight Microporous Coveralls with Elastic Wrists and Open Ankles
Shop All International Enviroguard Systems ProductsDescription
- Coveralls protect against particulates, light liquid splashes, and sprays in nonhazardous work environments
- Disposable coveralls provide a basic barrier against varnish, resins, fiberglass, and paints
- Keep productivity high and risk level low when wearing ValuGuard MP coveralls
- Secure, elastic wrists help keep sleeves in place to eliminate the possibility of skin exposure
- Standard zipper and set-in sleeves
- Environmentally friendly; can be recycled after use
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Microporous Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Standard |
| Ankle Style | Open |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.