Learn More
ValueMax™ Extra-Safe™ Lab Coats — Cranberry
Catalog No. 23900512A
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-900-512A Small
23-900-512B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-512A Supplier ValueMax™ Supplier No. 3660CYS
This item is not returnable. View return policy
Static-free—no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Specifications
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material SMS
Color Cranberry
Closure Type Snaps
Disposable Single-use
Style Knee-length
No. of Pockets 3
Latex Free Yes
Thickness 3-ply
Certifications/Compliance OSHA
Construction 3-ply SMS