Learn More
BURKLE INC Extension rod, 23.62 in. (60 cm), PTFE scoop
Supplier:  BURKLE INC 53890001
Encompass_Preferred
This extension rod complements the Bürkle PTFE dipper, allowing for sampling from greater distances. The rod is reinforced with a steel core and fully coated with PTFE to ensure maximum chemical resistance — especially useful when working with aggressive media.
Despite the added length, handling remains easy. For example, samples can be taken from large reactors, storage containers, or bodies of water without having to stand directly over the opening.
Catalog No. 50-000-26089
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.