Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-1+2+3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980872
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0872 Supplier Bioss Supplier No. BS10427R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Synapotobrevins, syntaxins, and the synaptosomal-associated protein SNAP25 are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. The protein encoded by this gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Mutations in this gene are associated with autosomal dominant spastic ataxia 1. Multiple alternative splice variants have been described, but the full-length nature of some variants has not been defined.

Specifications

Antigen VAMP-1+2+3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Vamp1
Gene Accession No. P23763
Gene Alias BOS_6013; DKFZp686H12131; lew; MRXSYP; SPAX1; Syb1; Syb-1; synaptobrevin 1; Synaptobrevin-1; VAMP1; VAMP-1; vesicle associated membrane protein 1; Vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1)
Gene Symbols Vamp1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human VAMP-1+2+3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.