Learn More
StatLab Van Gieson's Stain, 1 Pint
SDP
Supplier:  StatLab STVGIPT
Encompass_Preferred
When stained with the MasterTech Van Gieson’s Stain, collagen will stain pink or deep red, and cytoplasm will be stained yellow. Often times, Van Gieson's is used as a counterstain to other methods. In these instances, nuclear staining might not be necessary.
Catalog No. NC9953677
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.