Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-Molybdate Reagent for Phosphorus, Reagents

Vanadate-Molybdate Reagent for Phosphorus, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CV1020001A

Quantity 1 L

 View more versions of this product

Catalog No. 01-333-403


Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7647-01-0 , 12054-85-2
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Yellow
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.1
Non-sterile
Vanadate-Molybdate Reagent for Phosphorus
7732-18-5 , 7647-01-0 , 12054-85-2
H2O
O
18.02
Poly Bottle
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.