Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-Molybdate Reagent for Phosphorus, Reagents

Vanadate-Molybdate Reagent for Phosphorus, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$96.00 - $631.00

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 12054-85-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-333-404
Accessories View Documents
Reagents Holdings Llc
CV10200010F
10 L
Each for $631.00
Add to Cart
 
01-333-403
Accessories View Documents
Reagents Holdings Llc
CV1020001A
1 L
Each for $96.00
Add to Cart
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 7647-01-0 , 12054-85-2
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Yellow
100°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
1.1
Liquid
7732-18-5 , 7647-01-0 , 12054-85-2
H2O
O
18.02
Cubitainer
Non-sterile
Vanadate-Molybdate Reagent for Phosphorus
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.