Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadate-Molybdate Reagent for Phosphorus, Reagents

Catalog No. 01333404
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-404 10 L
01-333-403 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-404 Supplier Reagents Holdings Llc Supplier No. CV10200010F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanadate-Molybdate Reagent for Phosphorus, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 12054-85-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Yellow
Boiling Point 100°C
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.1
Quantity 10 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Vanadate-Molybdate Reagent for Phosphorus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.