Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(III) 2,4-pentanedionate, typically 97%

Catalog No. p-7049798
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2294014 25 g
AA2294022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2294014 Supplier Thermo Scientific Chemicals Supplier No. 02294014
Only null left
Add to Cart

CAS: 13476-99-8 | C15H21O6V | 348.27 g/mol

Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13476-99-8
Molecular Formula C15H21O6V
Molecular Weight (g/mol) 348.27
MDL Number MFCD00000033
InChI Key ZVFSRLXFKIWGRT-UHFFFAOYSA-N
Synonym Vanadium(III) acetylacetonate
PubChem CID 122198782
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;vanadium
SMILES [V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Quantity 25 g
UN Number UN3285
Sensitivity Air Sensitive
Solubility Information Soluble in water 2g/L,toluene,ethanol,acetone.
Formula Weight 348.27
Percent Purity 97% (typically)
Physical Form Powder
Chemical Name or Material Vanadium(III) 2,4-pentanedionate
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber 236-759-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.