Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(III) fluoride, 98%

Catalog No. AA8111509
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8111509 10 g
AA8111504 2 g
AA8111518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8111509 Supplier Thermo Scientific Chemicals Supplier No. 08111509
Only null left
Add to Cart

CAS: 10049-12-4 | F3V | 107.94 g/mol

Employed in oxygen-sensitive applications, such as metal production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-12-4
Molecular Formula F3V
Molecular Weight (g/mol) 107.94
MDL Number MFCD00042553
InChI Key PASVSMBJEYQYSN-UHFFFAOYSA-K
Synonym Vanadium trifluoride
IUPAC Name vanadium(3+) trifluoride
SMILES [F-].[F-].[F-].[V+3]

Specifications

Melting Point ≥1406°C
Physical Form Powder
Quantity 10 g
UN Number UN2923
Merck Index 14,9922
Solubility Information Practically insoluble in water, acetone, and glacial acetic acid
Formula Weight 107.94
Percent Purity 98%
Sensitivity Moisture sensitive
Density 3.363 g/mL at 19°C
Chemical Name or Material Vanadium (III) fluoride
Show More Show Less
Hazard Category H301-H314-H330-H335
Hazard Statement GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 233-169-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.