Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium(V) trichloride oxide, V+5 28.5% min

Catalog No. AA8111809
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8111809 10 g
AA8111836 500 g
AA8111822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8111809 Supplier Thermo Scientific Chemicals Supplier No. 08111809

CAS: 7727-18-6 | Cl3OV | 173.29 g/mol

Vanadium(V) trichloride oxide is used as a catalyst in olefin polymerization reaction, especially for the production of ethylene-propylene rubber. It is also used to synthesis organovanadium compounds and as a mordant in dyeing. It acts as a precursor to prepare vanadium pentoxide, vanadyl dichloride and dioxovanadium monochloride. Further, it serves as a reagent and a strong oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-18-6
Molecular Formula Cl3OV
Molecular Weight (g/mol) 173.29
MDL Number MFCD00011459
InChI Key WMZFXRASJAIALG-UHFFFAOYSA-K
Synonym vanadyl trichloride, trichlorooxovanadium, vanadium v trichloride oxide, trichlorooxovanadium oxide, vanadium v oxytrichloride, trichlorooxo vanadium, vanadium, trichlorooxo-, t-4, vanadium, trichlorooxo, vanadium oxide trichloride, vanadium trichloride oxide
PubChem CID 24410
IUPAC Name oxovanadium;trihydrochloride
SMILES [Cl-].[Cl-].[Cl-].O=[V+5]

Specifications

Boiling Point 126°C to 127°C
Melting Point -77°C
Color Yellow
Physical Form Liquid
Quantity 10 g
UN Number UN2443
Merck Index 14,9927
Solubility Information Miscible with benzene,dichloromethane and hexane.
Formula Weight 173.3
Concentration or Composition (by Analyte or Components) V+5 28.5% min
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 1.829 g/mL
Chemical Name or Material Vanadium (V) trichloride oxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: VANADIUM OXYTRICHLORIDE
EINECSNumber 231-780-2
RTECSNumber YW2975000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.