Learn More
Sigma Organic Chemistry Vanadium (V) Oxide Nanosheets | 10ML | 1314-6-2-1
Supplier: Sigma Organic Chemistry 92017710ML

Vanadium (V) Oxide Nanosheets, 10ml
About This Item:
Linear Formula: V2O5
Unspsc Code: 12352200
Storage: Room Temperature
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.