Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanado - Molybdate Reagent, AOAC, Ricca Chemical
SDP

Catalog No. 89851 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
89851 4 L Natural Poly Bottle
898516 500 mL Natural Poly Bottle
898532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89851 Supplier Ricca Chemical Company Supplier No. 89851
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Vanado - Molybdate Reagent
CAS 7732-18-5, 7601-90-3, 12054-85-2, 7803-55-6
Color Colorless to Yellow
CAS Min % 70.0, 26.29, 1.41, 0.07
CAS Max % 73.0, 27.37, 1.46, 0.07
Physical Form Liquid
Quantity 4 L
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Natural Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.