Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Vandetanib

Catalog No. AC464330010
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464330010 1 g
AC464332500 250 mg
AC464330050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC464330010 Supplier Thermo Scientific Chemicals Supplier No. 464330010
Add to Cart

CAS: 443913-73-3 | C22H24BrFN4O2 | 475.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443913-73-3
Molecular Formula C22H24BrFN4O2
Molecular Weight (g/mol) 475.36
InChI Key UHTHHESEBZOYNR-UHFFFAOYSA-N
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
SMILES COC1=C(OCC2CCN(C)CC2)C=C2N=CN=C(NC3=CC=C(Br)C=C3F)C2=C1

Specifications

Melting Point 240.0°C to 243.0°C
Solubility Soluble in methanol
Quantity 1 g
Formula Weight 475.35
Chemical Name or Material Vandetanib

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.