Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vang-like Protein 1/VANGL1 Antibody (497707) [Janelia Fluor™ 525], Novus Biologicals™
SDP

Catalog No. NB656076
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB656076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB656076 Supplier R&D Systems Supplier No. FAB5476JF525

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Vang-like Protein 1/VANGL1 Monoclonal antibody specifically detects Vang-like Protein 1/VANGL1 in Human samples. It is validated for Western Blot,Immunocytochemistry

Specifications

Antigen Vang-like Protein 1/VANGL1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 497707
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), vang-like 1 (van gogh, Drosophila), vang-like protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human Vang-like Protein 1/VANGL1, Glu242-Val524, Accession # Q8TAA9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 81839
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.