Learn More
Medchemexpress LLC Vanillic acid-13C6 | 201612-56-8 | MFCD00002551 | 99.9% | 174.10 | C2 13C6H8O4 | 25mg
SDP
Supplier:  Medchemexpress LLC HYW77822525MG
Encompass_Preferred
Vanillic acid-13C6 is the 13C labeled isotope of Vanillic acid-13C6(HY-N0708 ) Vanillic acid is a flavoring agent found in edible plants and fruits and in the root of angelica Vanillic acid inhibits NF- B activation It has anti-inflammatory and antimicrobial activities
Catalog No. 50-002-70244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.