Learn More
Medchemexpress LLC Vanillic acid glucos 5mg | 32142-31-7 | 330.29 | C14H18O9 | 5 MG
Supplier: Medchemexpress LLC HY1147605MG
Vanillic acid glucoside (vanillic acid 4-β-D-glucoside) is a plant-derived glucoside of vanillic acid supplied as a 5 mg research reagent for laboratory use. It is an off-white to light brown solid (C14H18O9, MW 330.29) typically used as a reference standard or for studies of plant metabolites and phytotoxic activity.
- High purity: 98.18% by supplier analysis.
- Compact 5 mg pack suitable for analytical and small-scale experiments.
- Off-white to light brown solid, easy to weigh and dissolve.
- Useful as a reference standard and for biochemical or phytochemical studies.
- Storage: keep sealed at -20°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.