Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Vannas Spring Scissors
SDP

Catalog No. 10000499 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
2mm, 0.05mm Straight
2mm, 0.05mm, Curved
2mm, 0.15mm Straight
2mm, 0.15mm, Curved
3mm tip, 0.2mm wide, curved
8.25cm, 0.1mm Tip, Straight
8.5cm, 0.3mm Tip, Bent
8.5cm, 0.3mm Tip, Side Angled
8.5cm, 0.3mm Tip, Slight Curve
8.5cm, 0.3mm Tip, Straight
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-499 8.5cm, 0.3mm Tip, Bent
10-000-496 8.5cm, 0.3mm Tip, Straight
10-000-497 8.5cm, 0.3mm Tip, Slight Curve
10-000-498 8.5cm, 0.3mm Tip, Side Angled
10-000-500 8.25cm, 0.1mm Tip, Straight
10-001-445 2mm, 0.15mm Straight
10-001-446 2mm, 0.15mm, Curved
10-001-452 3mm tip, 0.2mm wide, curved
10-001-455 2mm, 0.05mm Straight
10-001-456 2mm, 0.05mm, Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10-000-499 Supplier Stoelting™ Supplier No. 5213019P
Add to Cart
Add to Cart

Stoelting™ Vannas spring scissors are generally the smallest scissors available and are offered in several different scissor configurations

Specifications

For Use With (Application) Neuroscience
Model 8.5cm, 0.3mm Tip, Bent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.