Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Micro Ports
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- May require additional adapters to fit your container
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Exhaust Filters |
| Material | White Polypropylene (PP) |
| Size | NPT(M) x Luer Lock, Male |
| Thread Size (English) | 1/4 in. NPT(M) |
| For Use With (Application) | Solvent Safety |
| Type | Filter Adapter, Luer lock x 1/4 in. NPT(M), natural polypropylene; 1/ea |