Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(F)
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- May require additional adapters to fit your container
- Adjustable for flexible positioning
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Exhaust Filters |
| Material | Acetal |
| Size | NPT(M) x NPT(F), adjustable |
| Thread Size (English) | 1/4 in. NPT(M) x 1/4 in. NPT(F) |
| For Use With (Application) | Solvent Safety |
| Type | Adjustable Filter Adapter, 1/4 in. NPT(F), black acetal; 1/ea |
| Color | Black |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.