Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)

Catalog No. 13311188
Change view
Click to view available options
Size:
1/4 in. NPT(M) x 1/2 in. NPT(M)
1/4 in. NPT(M) x 1/4 in. NPT(M)
1/8 in. NPT(M) x 1/4 in. NPT(M)
45 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M)
90 elbow, 1/4 in. NPT(M) x 1/2 in. NPT(M)
90 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
13-311-188 1/4 in. NPT(M) x 1/4 in. NPT(M)
13-311-189 1/4 in. NPT(M) x 1/2 in. NPT(M)
13-311-191 90 elbow, 1/4 in. NPT(M) x 1/2 in. NPT(M)
13-311-195 45 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M)
13-311-196 90 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M)
13-311-202 1/8 in. NPT(M) x 1/4 in. NPT(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-311-188 Supplier Cole-Parmer™ Supplier No. 1201986
Add to Cart
Add to Cart

VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources

  • Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
  • May require additional adapters to fit your container

Warranty and Services

None

Specifications

For Use With (Equipment) VapLock Exhaust Filters
Material Polypropylene
Size 1/4 in. NPT(M) x 1/4 in. NPT(M)
Thread Size (English) 1/4 in. NPT(M)
For Use With (Application) Solvent Safety
Type Filter Adapter; 1/ea
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.