Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Quick-Disconnect
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- Features Justrite Centura quick disconnect
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Exhaust Filters |
| Material | Stainless Steel, PTFE |
| Size | NPT(M) x Quick Disconnect |
| Thread Size (English) | 1/4 in. NPT(M) |
| For Use With (Application) | Solvent Safety |
| Type | Filter Adapter |
| Quantity | 1 Ea. |
| Color | Gray |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.