Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Adapters for Flexible Tubing

Catalog No. 13311232
Change view
Click to view available options
Size:
1/16 in. Barb x NPT(M)
1/2 in. Barb x NPT(M)
1/4 in. Barb x NPT(M)
1/8 in. Barb x NPT(M)
3/16 in. Barb x NPT(M)
3/4 in. Barb x NPT(M)
3/8 in. Barb x NPT(M)
5/16 in. Barb x NPT(M)
5/32 in. Barb x NPT(M)
5/8 in. Barb x NPT(M)
Thread Size (English):
1/2 in. NPT(M)
1/4 in. NPT(M)
1/8 in. NPT(M)
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Size (English)
13-311-232 3/8 in. Barb x NPT(M) 1/8 in. NPT(M)
13-311-220 1/8 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-222 1/4 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-223 5/16 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-224 3/8 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-225 1/2 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-226 5/32 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-228 1/8 in. Barb x NPT(M) 1/8 in. NPT(M)
13-311-230 1/4 in. Barb x NPT(M) 1/8 in. NPT(M)
13-311-231 5/16 in. Barb x NPT(M) 1/8 in. NPT(M)
13-311-233 1/4 in. Barb x NPT(M) 1/2 in. NPT(M)
13-311-234 3/8 in. Barb x NPT(M) 1/2 in. NPT(M)
13-311-236 5/8 in. Barb x NPT(M) 1/2 in. NPT(M)
13-311-237 3/4 in. Barb x NPT(M) 1/2 in. NPT(M)
13-311-221 3/16 in. Barb x NPT(M) 1/4 in. NPT(M)
13-311-227 1/16 in. Barb x NPT(M) 1/8 in. NPT(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 13-311-232 Supplier Cole-Parmer™ Supplier No. 1202030
Add to Cart
Add to Cart

VapLock™ Adapters for Flexible Tubing are designed for secure leak-resistant connections

Use with flexible, soft-wall tubing

Warranty and Services

None

Specifications

For Use With (Equipment) VapLock Solvent Containment Systems
Material Polypropylene
Size 3/8 in. Barb x NPT(M)
Thread Size (English) 1/8 in. NPT(M)
For Use With (Application) Solvent Safety
Type Tubing Adapter, 3/8 in. ID x 1/8 in. NPT(M), Polypropylene, 10/Pk.
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.