Learn More
Cole-Parmer™ VapLock™ Adapters with Large Bore for Semi-Rigid Tubing
Catalog No. 13311241
Change view
Click to view available options
Size:
1/2 in. OD Compression x NPT(M)
3/8 in. OD Compression x NPT(M)
5/16 in. OD Compression x NPT(M)
Thread Size (English):
1/2 in. NPT(M)
1/4 in. NPT(M)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thread Size (English)
13-311-241 1/2 in. OD Compression x NPT(M) 1/4 in. NPT(M)
13-311-239 5/16 in. OD Compression x NPT(M) 1/4 in. NPT(M)
13-311-240 3/8 in. OD Compression x NPT(M) 1/4 in. NPT(M)
13-311-242 3/8 in. OD Compression x NPT(M) 1/2 in. NPT(M)
13-311-243 1/2 in. OD Compression x NPT(M) 1/2 in. NPT(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-311-241 Supplier Cole-Parmer™ Supplier No. 1202039
Add to Cart
Add to Cart
VapLock™ Adapters with Large Bore for Semi-Rigid Tubing are designed for secure leak-resistant connections
Integral ferrule prevents loss of small parts
Warranty and Services
None
Specifications
For Use With (Equipment) VapLock Solvent Containment Systems
Material Polypropylene
Size 1/2 in. OD Compression x NPT(M)
Thread Size (English) 1/4 in. NPT(M)
For Use With (Application) Solvent Safety
Type Tubing Adapter, 1/2 in. OD tubing x 1/4 in. NPT(M), Polypropylene; 5/Pk.