Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Adapters for Semi-Rigid Tubing, UNF(M)

Catalog No. 13311206
Change view
Click to view available options
Size:
1/16 in. OD Compression x UNF(M)
1/8 in. OD Compression x UNF(M)
2 mm OD Compression x UNF(M)
2.2 mm OD Compression x UNF(M)
2.5 mm OD Compression x UNF(M)
3 mm OD Compression x UNF(M)
3/16 in. OD Compression x UNF(M)
Material:
Black / White PP / PTFE
Blue / Yellow PP / ETFE
Green / Blue PP / ETFE
Orange / White PP / ETFE
Red / Green PP / ETFE
White / Blue POM / ETFE
Yellow / Orange PP / ETFE
Thread Size (English):
1/4 in.-28 UNF(M)
5/16 in.-24 UNF(M)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Thread Size (English)
13-311-206 2 mm OD Compression x UNF(M) Red / Green PP / ETFE 1/4 in.-28 UNF(M)
13-311-204 3/16 in. OD Compression x UNF(M) White / Blue POM / ETFE 5/16 in.-24 UNF(M)
13-311-205 1/16 in. OD Compression x UNF(M) Green / Blue PP / ETFE 1/4 in.-28 UNF(M)
13-311-207 2.2 mm OD Compression x UNF(M) Black / White PP / PTFE 1/4 in.-28 UNF(M)
13-311-208 2.5 mm OD Compression x UNF(M) Orange / White PP / ETFE 1/4 in.-28 UNF(M)
13-311-209 3 mm OD Compression x UNF(M) Yellow / Orange PP / ETFE 1/4 in.-28 UNF(M)
13-311-210 1/8 in. OD Compression x UNF(M) Blue / Yellow PP / ETFE 1/4 in.-28 UNF(M)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-311-206 Supplier Cole-Parmer™ Supplier No. 1202004
Add to Cart
Add to Cart

VapLock™ Adapters for Semi-Rigid Tubing, UNF(M) are designed for secure liquid-tight connections

  • Color-coded for easy identification
  • Features two-piece nut and ferrule system

Warranty and Services

None

Specifications

For Use With (Equipment) VapLock Solvent Containment Systems
Material Red / Green PP / ETFE
Size 2 mm OD Compression x UNF(M)
Thread Size (English) 1/4 in.-28 UNF(M)
For Use With (Application) Solvent Safety
Type Tubing Adapter, 1.8-2.0 mm OD x 1/4-28 UNF(M), red PP w/ green ETFE; 10/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.