Learn More
Cole-Parmer™ VapLock™ Breakthrough Color Indicator for Exhaust Filters
Description
- Changes color when exposed to organic solvent vapors
- Optically clear acrylic housing for easy view
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Exhaust Filters |
| Includes | 5/Pk. |
| Material | Acrylic |
| For Use With (Application) | Solvent Safety |
| Type | Breakthrough Indicator for Exhaust Filters |