Description
- Minimize evaporation and relieve pressure to improve safety
- Allow air into supply container to displace vacuum and keep solvent flowing
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Solvent Containment Systems |
| Includes | 1 Ea. |
| Material | PTFE, FFKM |
| Thread Size | 1/4 in.-28 UNF(M) |
| For Use With (Application) | Solvent Safety |
| Product Type | Inlet Check Valve; 0.1-0.25 psi crack pressure |