Learn More
Cole-Parmer™ VapLock™ Reversible Valves for Solvent Management
Description
- Minimize evaporation and relieve pressure to improve safety
- Change orientation to switch between inlet and outlet flow regulation
Warranty and Services
None
Specifications
Specifications
| For Use With (Equipment) | VapLock Solvent Containment Systems |
| Material | Polypropylene, EPDM |
| Pressure | 0.1 to 0.25 psi crack pressure |
| Thread Size (English) | 2 x 1/4 in. NPT(M) x 1/4 in. NPT(M) |
| For Use With (Application) | Solvent Safety |
| Type | Reversible Valve |