Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI Cap Size and 20-L Container

Catalog No. 13311269 Shop All Oakton Products
Change view
Click to view available options
Material:
PTFE Manifold
Polypropylene Manifold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
13-311-269 Polypropylene Manifold
13-311-270 PTFE Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-311-269 Supplier Cole-Parmer™ Supplier No. 1202067
Add to Cart
Add to Cart

VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI cap size and 20-L Container are a complete solution to safely collect solvent waste from laboratory instruments

Collect solvent waste from up to 9 effluent streams Minimize exposure to harmful chemical vapors

Warranty and Services

None

Specifications

Description 70 mm Rieke 6 TPI, Polypropylene manifold, 20 L Polyethylene drum
For Use With (Equipment) Chromatography Systems
Type Complete Solvent Collection System
Solvent Channels 9
Material Polypropylene Manifold
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.