Learn More
Cole-Parmer™ VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container
Catalog No. 13311271 Shop All Oakton Products
Change view
Click to view available options
Material:
PTFE
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
13-311-271 Polypropylene
13-311-272 PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-311-271 Supplier Cole-Parmer™ Supplier No. 1202069
Add to Cart
Add to Cart
VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container are a complete solution to safely collect solvent waste from laboratory instruments
  • FM-approved container offers added safety
  • Collect solvent waste from up to 10 effluent streams
  • Minimize exposure to harmful chemical vapors
Warranty and Services
None
Specifications
For Use With (Application) Solvent Waste Collection
Description Quick disconnect, Polypropylene manifold, 20 L FM-approved PE drum
For Use With (Equipment) Chromatography Systems
Type Complete Solvent Collection System
Material Polypropylene
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov