Learn More
Cole-Parmer™ VapLock™ Solvent Collection Systems with Quick Disconnects and FM-Approved Container
Shop All Oakton ProductsDescription
- FM-approved container offers added safety
- Collect solvent waste from up to 10 effluent streams
- Minimize exposure to harmful chemical vapors
Warranty and Services
None
Specifications
Specifications
| For Use With (Application) | Solvent Waste Collection |
| Description | Quick disconnect, PTFE manifold, 20 L FM-approved PE drum |
| For Use With (Equipment) | Chromatography Systems |
| Type | Complete Solvent Collection System |
| Material | PTFE |