Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex™ VapLock™ Solvent Delivery Cap Kits

Catalog No. 1202091
Change view
Click to view available options
Type:
Solvent Delivery Cap Kit, Normal Phase
Solvent Delivery Cap Kit, Reverse Phase
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
12-020-91 Solvent Delivery Cap Kit, Reverse Phase
12-020-92 Solvent Delivery Cap Kit, Normal Phase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-020-91 Supplier Spex™ Supplier No. 1202091
Add to Cart
Add to Cart

Reduce chemical evaporation and exposure to hazardous vapors on multiple carboys

  • Less expensive than ordering individual components
  • Color coded caps allow easy solvent identification
  • Choose from FFKM, EPDM, or Viton™ valve seal options
  • Ideal for quaternary systems
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4''-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable. Caps with integrated air inlet valve and filter permit air to enter the bottle for vacuum displacement while solvent is pumped to the instrument. Valves are made of PTFE and 300-series SS components and feature a 50 μm PTFE filter to reduce particulate contamination.

Specifications

Includes Four color-coded GL45 safety caps in blue, red, yellow, and green, with eight connections for 1/8 in. OD tubing, one connection for 2.2 mm OD tubing, and four PTFE plugs.
Material EPDM
Thread Size GL45
Type Solvent Delivery Cap Kit, Reverse Phase
Color Green
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.