Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Solvent Delivery Caps

Catalog No. 13311017
Change view
Click to view available options
Size:
GL25
GL32
GL38
GL45
Nalgene 38-430
Wako 38 mm
Material:
PTFE, Polypropylene
PTFE, Polypropylene, Stainless Steel
Number of Channels:
Four Ports
Three Ports
Two Ports
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Number of Channels
13-311-017 GL38 PTFE, Polypropylene Four Ports
13-311-003 GL45 PTFE, Polypropylene Two Ports
13-311-025 Nalgene 38-430 PTFE, Polypropylene, Stainless Steel Three Ports
13-311-037 Wako 38 mm PTFE, Polypropylene, Stainless Steel Three Ports
13-311-052 GL32 PTFE, Polypropylene Three Ports
13-311-004 GL45 PTFE, Polypropylene Three Ports
13-311-050 GL32 PTFE, Polypropylene, Stainless Steel Four Ports
13-311-028 Nalgene 38-430 PTFE, Polypropylene Three Ports
13-311-013 GL38 PTFE, Polypropylene, Stainless Steel Three Ports
13-311-015 GL38 PTFE, Polypropylene Two Ports
13-311-051 GL32 PTFE, Polypropylene Two Ports
13-311-005 GL45 PTFE, Polypropylene Four Ports
13-311-024 Nalgene 38-430 PTFE, Polypropylene, Stainless Steel Two Ports
13-311-027 Nalgene 38-430 PTFE, Polypropylene Two Ports
13-311-041 Wako 38 mm PTFE, Polypropylene Four Ports
13-311-038 Wako 38 mm PTFE, Polypropylene, Stainless Steel Four Ports
13-311-000 GL45 PTFE, Polypropylene, Stainless Steel Two Ports
13-311-053 GL32 PTFE, Polypropylene Four Ports
13-311-048 GL32 PTFE, Polypropylene, Stainless Steel Two Ports
13-311-055 GL25 PTFE, Polypropylene Three Ports
13-311-012 GL38 PTFE, Polypropylene, Stainless Steel Two Ports
13-311-001 GL45 PTFE, Polypropylene, Stainless Steel Three Ports
13-311-039 Wako 38 mm PTFE, Polypropylene Two Ports
13-311-026 Nalgene 38-430 PTFE, Polypropylene, Stainless Steel Four Ports
13-311-036 Wako 38 mm PTFE, Polypropylene, Stainless Steel Two Ports
13-311-016 GL38 PTFE, Polypropylene Three Ports
13-311-040 Wako 38 mm PTFE, Polypropylene Three Ports
13-311-029 Nalgene 38-430 PTFE, Polypropylene Four Ports
13-311-054 GL25 PTFE, Polypropylene Two Ports
13-311-002 GL45 PTFE, Polypropylene, Stainless Steel Four Ports
13-311-014 GL38 PTFE, Polypropylene, Stainless Steel Four Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. 13-311-017 Supplier Cole-Parmer™ Supplier No. 1201817
Add to Cart
Add to Cart

VapLock™ Solvent Delivery Caps reduce chemical evaporation and exposure to hazardous vapors

Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD

Warranty and Services

None

Specifications

For Use With (Equipment) Solvent-Compatible Containers
Material PTFE, Polypropylene
Number of Channels Four Ports
Size GL38
Thread Size 1/4 in.-28
Thread Size (English) 1/4 in.-28 UNF(F)
For Use With (Application) Solvent Safety
Type Delivery Cap
Color Black/White
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.